Learn More
Eppendorf™ Platforms for New Brunswick™ S41i Incubator Shaker
Interchangeable, dedicated platform for the S41i.
Brand:  Eppendorf™ M1334-9927
 View more versions of this product
Product Code. 13498686
  • 863.44 EUR / Each
In Stock
to see stock.
Explore more special offers
Description
  • Dedicated platform
  • Interchangeable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4 L Erlenmeyer Flasks
New Brunswick™ S41i
3