Learn More
Platinum, nominally 40% on carbon black, Thermo Scientific Chemicals
463.00 EUR - 110505.00 EUR
Description
Platinum black commonly acts as a catalyst in chemical reactions, as a catalytic converter in automobiles and in the petroleum industry as a catalyst. It also vigorously catalyzes the decomposition of hydrogen peroxide into water and oxygen. It is used extensively as a laboratory equipment, electrical contacts, electrodes, platinum resistance thermometers, dentistry equipment, and jewelry. Compounds containing platinum (cisplatin, oxaliplatin and carboplatin) are applied in chemotherapy against certain types of cancer and also applied as pollution control catalysts for gasoline and diesel vehicles and in control of emissions of organics and nitrous oxides from industrial installations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-6-6 | |
MFCD00011179 |
Specifications
7440-6-6 | |
MFCD00011179 | |
Powder |
nominally 40% on carbon black | |
Soluble in aqua regia,but insoluble in hydrochloric and nitric acids. | |
Platinum |
Safety and Handling
P201-P202-P210-P240-P241-P281-P308+P313-P370+P378q-P501c
H228-H351
missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: II
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only