missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SI Analytics™ Plug Cable Combination LS 3 ANN
SI Analytics™ Plug Cable Combination LS 3 ANN is an electrode plug designed to be used with a SMEK plug head.
Brand: SI Analytics™ 285122715
This item is not returnable.
View return policy
Description
- Plug and cable combination
- 3m cable
- Includes 2 x 4mm banana plug
Specifications
Electrode Plug | |
LS 3 ANN | |
3m Cable, BNC Instrument Plug, + 2 x 4mm Banana Plug | |
3 m |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction