Learn More
Memmert™ Plug-In Tube Extension for UF750plus, Straight
Product Code. 17579575 Shop All Memmert Products
Click to view available options
Diameter (Metric):
O.D. 60.3 mm, I.D. 57 mm
Unit Size:
Each
Product Code. 17579575
Brand: Memmert™ UF750plusB29718
Only null left
Add to basket
Add to basket
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
  • Accessory
  • Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
  • Straight
Specifications
For Use With (Equipment) UF750plus Oven
Type Plug-in Tube Extension
Diameter (Metric) O.D. 60.3 mm, I.D. 57 mm