Learn More
Sampling Systems™ Pocket Sampler
Shop All Sampling Systems Products
Click to view available options
Length (Metric):
300 mm
600 mm
900 mm
1500 mm
2000 mm
Capacity (Metric):
10 mL
15 mL
20 mL
90 mL
Diameter (Metric):
25 mm
40 mm
Handle Material:
PTFE Inner Rod
Stainless Steel Inner Rod
missing translation for 'unitSize'
Each
Descrizione
- Made from high quality 316L stainless steel with PTFE or 316L Stainless Steel inner core
- For taking multi-level, discrete samples
- Suitable for free flowing powders
- Bayonet handle that allows for simple operation
- Sealing sleeve enables one pocket to be emptied at a time
- Samplers are supplied with pointed tips as standard
Specifica
Specifica
| Product Type | Sampler |
| Material | Stainless Steel (316L) |
| Includes | 4 Pockets |
| Handle Material | Stainless Steel Inner Rod |
| Capacity (Metric) | 90 mL |
| Diameter (Metric) | 40 mm |
| Length (Metric) | 2000 mm |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto