Learn More
SCHOTT AG Lighting and Imaging™ Polarization Filter Set for Ring Light
This Polarization Filter Set for Ring Light provides contrast with polarized light by using a polarizing and analyzing filter within the beam path. Assists in providing contrast with Stereo Microscopy to help identify inner structure materials or metal particles within others.
Brand:  SCHOTT AG Lighting and Imaging™ 158440
 View more versions of this product
Product Code. 10359600
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
  • Polarization Filter Set for fiber optic Ring Light.
  • Fits inside diameters of 58 or 66 mm.
  • Includes polarizer and analyzer.
  • For use with fiber optic Ring Light.
Specifications
Specifications
Filter
Stereo Microscopy Illumination
Polarizer and analyzer
58 mm
Fiber Optic Ringlight