missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly-L-Lysine Hydrobromide [MW 4,000-15,000], MP Biomedicals
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Specifications
Specifications
Quantity | 100 mg |
Formula Weight | 4,000 to 15,000 |
Chemical Name or Material | Poly-L-Lysine Hydrobromide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'