Learn More
3M™ Polycarbonate Lens Safety Goggles
Click to view available options
Lens Tint:
Amber
Specifications
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Amber |
Product Type | Safety goggles |
For Use With (Application) | Drilling, grinding and cleaning, stone size, Metallurgie |
Frame Color | Black |
No. per Pack | 1 Ea. |
Material | Polycarbonate |
UV Protection | Yes |