Learn More
Lenz Laborglasinstrumente™ Polyester-Glass Compound Clamps
548.00 EUR - 1055.00 EUR
Specifications
Quantity 1/Pk.
Material Polyester-glass Compound (Clamp), Steel (Rod)
For Use With (Equipment) For Fixing Lids Onto Reaction Vessels And Mounting On A Stand
View More Specs
Products 2
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
10814541
View Documents
Lenz Laborglasinstrumente™
06246515
150 Laboratory Clamp
1055.00 EUR
Each
11556814
View Documents
Lenz Laborglasinstrumente™
06246506
60 Laboratory Clamp
548.00 EUR
Each
Specifications
1/Pk.
Polyester-glass Compound (Clamp), Steel (Rod)
For Fixing Lids Onto Reaction Vessels And Mounting On A Stand