Learn More
Macherey-Nagel™ Polyester Sheets Polygram SIL G/UV254 TLC Micro-Set F3
Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
4 x 8 cm
Unit Size:
Each
Description
TLC Micro-Set F3 is the advanced set 3 for science education. It contains all chemicals required for - Separation of analgesics (pain relievers) and drug analysis based on the example of cinchona bark. For the separations with Micro-Set F3, the materials of TLC Micro-Set M is prerequisite.
Specifications
Specifications
| Material | Polyester Sheets Polygram SIL G |
| Quantity | 1 Ea. |
| Type | TLC Micro-Set F3 |
| Fluorescent Indicator | UV254 |
| Includes | 1 manual, 50 glass capillaries 1 μL; 50 polyester sheets Polygram SIL G/UV254, 4 x 8 cm; 5 Aspirin tablets; 5 Thomapyrin tablets; 20 folded filters MN 615 1/4, 11 cm diameter; 3 sample vials 8 mL (for Aspirin sample, Thomapyrin sample and cinchona bark extract); 5 g cinchona bark; 100 mL each of ethanol, 2-propanol, toluene - diethyl ether (61:39, v/v)*, spray reagent for caffeine (in aqueous acetone solution)* and spray reagent according to Dragendorff-Munier (aq. soln.); 50 mLea. of iron(III) chloride solution* and potassium hexacyanoferrate(III) solution (each aq. soln.); 30 mL ethyl acetate*; 25 mL each of 12.5 % ammonia and diethylamine*; 8 mL each of caffeine, paracetamol, quinine reference solutions (each in ethanol)* |
| Dimensions (L x W) | 4 x 8 cm |