Learn More
Hanna Instruments™ HI-1332P pH Electrode
The HI-1332P is a gel filled, plastic body pH electrode with double reference junction that can be used with most hand-held pH/mV meters. Ideal for use with meters that have CAL Check.
Brand: Hanna Instruments™ HI1332P
This item is not returnable.
View return policy
Description
- BNC connector and pin
- Gel filled
- Suitable for most hand-held pH/mV meters
- Ideal for use with meters that have CAL Check
- Ideal for chemicals, field applications, QC testing
Specifications
BNC | |
PolyEther-Imide | |
20°C to 40°C |
Ceramic | |
0 to 13 | |
pH Electrode |