Learn More
Kartell™ Plastilab™ Polyethylene Flat-Bottom Sample Vials
Click to view available options
Capacity: 25mL
Description
- Convenient push-on attached hinged lids
- Lightweight and unbreakable
- Translucent
- Suitable for storing food stuff
Specifications
Specifications
Capacity (Metric) | 25 mL |
Includes | Push on hinged lid |
Material | Polyethylene |
Thickness (Metric) | 1.6mm |
Type | Flat Bottom Sample Vial |
Closure Type | Push-on Hinged Lid |