missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene glycol, MP Biomedicals™
Click to view available options
Quantity:
1 kg
Specifications
Specifications
Color | White |
CAS | 25322-68-3 |
Physical Form | Powder |
Molecular Formula | (C2H4O)n |
MDL Number | MFCD01779601 |
Density | 1.1 to 1.2g/mL |
Synonym | Carbowax, Alkapol PEG-200 |
Quantity | 1 kg |
InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
SMILES | [H]OCCO |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'