Learn More
Buerkle™ Polyethylene Round All Purpose Boxes
Box for storing, packing, taking samples, sampling and transport of pastes, granules, powders, soil samples and frozen goods (-40 °C up to briefly +95 °C). Ideal use of space, can be stacked. Complete with press-in lid
4.25 EUR - 4.80 EUR
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10286363
|
Buerkle™
6210-0250 |
250 mL |
4.25 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10780773
|
Buerkle™
6210-0500 |
500 mL |
4.80 EUR
Each |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
Description
- PE, transparent
- No crevices
- |PE, transparent
Specifications
Transparent | |
Polyethylene | |
Round |