Learn More
Brady™ Polyethylene Tags for M611, BMP61 and BMP71
Multi-purpose tags for Thermal Transfer Portable Printers that can be used for a variety of applications including identification of multi-conductor cables, inventory, equipment, lockout, safety warning repair and work-in-progress.
Brand:  Brady™ 018347
 View more versions of this product
Product Code. 11552765
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Cross-laminated polyethylene provides extreme tear resistance and excellent cold-weather performance
  • TLS2200™ parts can also be printed on BMP™71 Label Printer provided roll adapter (M71-ADAPT) is used
Specifications
Specifications
Not Autoclavable
White
Polyethylene (B-109)
Non-sterile
BMP™61, BMP™71, and BradyPrinter M611 Mobile Label Printers, TLS 2200™ and TLS-PC LINK™ Thermal Labeling Systems
100 Labels
-40°C to +49°C
76.2 mm
12.5 mm
57.9 mm
Non-adhesive
Matte
Thermal Transfer
Data and Telecommunications Labeling, Inventory Labeling, Lockout Tagout, Safety Identification
1
Rectangle
0.22 mm
19.1 mm
76.2 mm