Learn More
Fisherbrand™ Polypropylene 96 Well Microplate
Click to view available options
Total Volume (Metric) Well:
2 mL
Unit Size:
Pack of 50
Description
- Unique, patented sealing cap allows penetration of a needle through the cap into each well, minimising coring of the needle as the base of each well of the cap has been reduced in thickness
- Small radius on all corners to prevent sample precipitation and improve concentration
- DNase and RNase free
- Applications include SPE, HPLC, MS, liquid handling, automation, robotics
Specifications
Specifications
| Format | Standard |
| Array | 8 x 12 |
| Material | Polypropylene |
| No. of Wells | 96 |
| Total Volume (Metric) Well | 2 mL |
| Well Shape | Square, Conical Bottom (V-well) |
| Dnase Free | Yes |
| No. per Pack | 50 |
| RNase Free | Yes |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Polypropylene 96 Well Microplate
Spot an opportunity for improvement?Share a Content Correction