missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ Polypropylene Bottle

Brand:  Kautex™ 2000086458

 View more versions of this product

Product Code. 11301994

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Low weight, leakproof and good transparency and glaze
  • Outstanding chemical resistance and thermal stability
  • Balanced stiffness and impact resistance
  • Easily recyclable
  • Ideal for product presentation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

500 mL
28.6 mm
Polypropylene
1 Ea.
1 Ea.
Yes
Narrow
Bottle
75 mm
Transparent
Packaging, Sending Samples, Presentation and Transport
135/Cs.
Bottle
Yes
Autoclavable
169 mm
32 mm
Screw Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers