missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ Polypropylene Bottle

Brand:  Kautex™ 2000086457

 View more versions of this product

Product Code. 11391984

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Low weight, leakproof and good transparency and glaze
  • Outstanding chemical resistance and thermal stability
  • Balanced stiffness and impact resistance
  • Easily recyclable
  • Ideal for product presentation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

250 mL
18.6 mm
Polypropylene
1 Ea.
1 Ea.
Yes
Narrow
Bottle
60 mm
Transparent
Packaging, Sending Samples, Presentation and Transport
247/Cs.
Bottle
Yes
Autoclavable
136 mm
32 mm
Screw Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.