Learn More
Vitlab™ Polypropylene Bucket with Spout
Click to view available options
Capacity (Metric):
15 L
Unit Size:
Each
Description
- Transparent
- Divided into 1 liter segments
- Stable handle and spout for easy emptying
- Highly resistant to chemicals
- Without lid
Specifications
Specifications
| Product Type | Bucket |
| Capacity (Metric) | 15 L |
| Graduations | Yes |
| Material | Polypropylene |
| Color | Transparent |
| Diameter (Metric) Outer | 310 mm |
| Height (Metric) | 370 mm |
| Quantity | 1 Ea. |