Få mere at vide
Få mere at vide
Fisherbrand™ Polypropylene Natural Pipet Tip
Shop alle Fisherbrand produkter
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
1 to 200 μL
1 to 250 μL
1 to 300 μL
200 to 1000 μL
250 to 1000 μL
500 to 2500 μL
Förpackningsstorlek
Pack of 1000
Pack of 250
Beskrivelse
- Made of high quality polypropylene
- Tight fit due to the precisely fitting cone
- Optimal handling due to the stable shape
- Autoclavable
- Guaranteed dust-free due to special ionization treatment
- Available pre-sterilized or non-sterile
- Excellent transparency due to high quality surfaces
- Minimal residual wetting due to controlled quality of raw materials
- Excellent volume accuracy due to precise tip orifice
- Practice-oriented tip volumes
- Aeroject™ tips with special filters made of hydrophobic PE-HD as
- Aerosol barrier and for protection against contamination
- Suitable for all commonly used pipet types
Tekniske data
Tekniske data
| Volume (Metric) | 1 to 200 μL |
| Color | Natural |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion