Learn More
Fisherbrand™ Polypropylene Natural Pipet Tip
Product Code. 11597452 Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
1 to 200 μL
1 to 250 μL
1 to 300 μL
200 to 1000 μL
250 to 1000 μL
500 to 2500 μL
Unit Size:
Pack of 1000
Pack of 250
Product Code. 11597452
Make pipetting simple and safer for even the most sensitive applications. Fisherbrand™ Polypropylene Natural Pipet Tips are perfectly fit for manual or electronic, single or multichannel pipets.
  • Made of high quality polypropylene
  • Tight fit due to the precisely fitting cone
  • Optimal handling due to the stable shape
  • Autoclavable
  • Guaranteed dust-free due to special ionization treatment
  • Available pre-sterilized or non-sterile
  • Excellent transparency due to high quality surfaces
  • Minimal residual wetting due to controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice
  • Practice-oriented tip volumes
  • Aeroject™ tips with special filters made of hydrophobic PE-HD as
  • Aerosol barrier and for protection against contamination
  • Suitable for all commonly used pipet types
Specifications
Volume (Metric) 250 to 1000 μL
Color Natural