Learn More
Eppendorf™ PCR Tubes - Singles and Strips
Eppendorf PCR Tubes ensure uniform heat distribution to the sample for optimal PCR performance. The user-friendly design allows for easy opening and provides a secure seal to prevent evaporation.
Brand: Eppendorf™ 0030124359
This item is not returnable.
View return policy
Description
Ensure efficient and homogenous heat transfer with these thin-walled polypropylene tubes. Eppendorf™ Polypropylene PCR Tubes are easy to open, but provide tight sealing to prevent evaporation in PCR.
- Design ensures an airtight seal to prevent contamination and evaboration.
- Thin polypropylene walls ensure efficient and homogeneous heat transfer or fast and high-quality PCR results.
- 8 reaction tubes in strip-format – ideal for small sample volumes. Easily adaptable for automation.
- Strips are sealable using flat or domed strip-lids.
- PCR Clean: Certified free from human DNA, DNase, RNase and PCR inhibitors).
Specifications
Transparent | |
960 Pk. | |
PCR | |
Polypropylene | |
Snap Cap | |
PCR Tubes (8-tube Strip) |
0.2 mL | |
Polypropylene | |
Non-sterile | |
PCR Clean (Non-sterile) | |
8-Tube Strip |