Learn More
Eppendorf™ PCR Tubes - Singles and Strips
Shop All Eppendorf Products
Click to view available options
Closure Type:
Domed Strip Cap
Flat Strip Cap
Hinged Cap
Not Included
Format:
8-tube Strips
Fast PCR Tube Strips
Single Tube
Purity or Quality Grade:
Forensic DNA Grade
PCR Clean (Non-sterile)
Volume (Metric):
0.1 mL
0.2 mL
0.5 mL
Unit Size:
Pack of 1000
Pack of 120
Pack of 500
Description
Ensure efficient and homogenous heat transfer with these thin-walled polypropylene tubes. Eppendorf™ Polypropylene PCR Tubes are easy to open, but provide tight sealing to prevent evaporation in PCR.
- Design ensures an airtight seal to prevent contamination and evaboration.
- Thin polypropylene walls ensure efficient and homogeneous heat transfer or fast and high-quality PCR results.
- 8 reaction tubes in strip-format – ideal for small sample volumes. Easily adaptable for automation.
- Strips are sealable using flat or domed strip-lids.
- PCR Clean: Certified free from human DNA, DNase, RNase and PCR inhibitors).
Specifications
Specifications
| Volume (Metric) | 0.1 mL |
| Sterility | Non-sterile |
| Material | Polypropylene |
| Purity or Quality Grade | PCR Clean (Non-sterile) |
| Closure Type | Flat Strip Cap |
| Autoclavable | Autoclavable |
| Closure Included | Yes |
| Format | Fast PCR Tube Strips |
| Product Type | PCR Tube Strip |