missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ Polypropylene Reagent Bottle
Click to view available options
Capacity (Metric):
500 mL
Diameter (Metric):
87 mm
Height (Metric):
172 mm
Description
- Transparent
- With screw cap made of polypropylene
- Good chemical resistance, ideal for long-term storage of liquids Autoclavable at 121 °C (2 bar) according to DIN EN 285
- Wider mouth for simple filling
- Ideal for powders and paste-like materials
Specifications
Specifications
Capacity (Metric) | 500 mL |
Color | Transparent |
Includes | Screw cap |
Material | Polypropylene |
No. per Pack | 10/Pk. |
Certifications/Compliance | DIN EN 285 |
Autoclavable | Autoclavable |
Height (Metric) | 172 mm |
Closure Material | Polypropylene |
Diameter (Metric) | 87 mm |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction