Learn More
VITLAB™ Polypropylene Scoops
Sample, measure and weigh small volumes. VITLAB™ Polypropylene Scoops are durable, chemically resistant shovels designed for precise measurement. Molded of autoclavable polypropylene with a precision-formed filling edge, flat bottom and comfortable handle. Available in several sizes.
Brand:  VITLAB™ 39294
 View more versions of this product
Product Code. 11867920
  • 5.80 EUR / Pack of 12
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Description
These molded one-piece laboratory scoops are made of FDA-grade polypropylene, well suited for sampling, measuring and weighing needs. The shape and size are great for taking samples directly from bottles or containers.
  • Constructed of sturdy, white polypropylene
    • Heat resistant and autoclavable
    • High chemical resistance
    • Recyclable and reusable
  • Designed for precise, easy measurement
    • Molded with a precision-formed filling edge
    • Comfortable, stable handle with easily readable volume on the upper side
    • Flat-bottom design stays upright when placed on a flat surface
  • Well suited for a variety of lab applications
    • Food, pharmaceutical and environmental sampling
    • Chemical and material measuring
    • For use as a weighing boat
Specifications
Specifications
Measuring Scoop
Autoclavable
5 mL
White
Sampling, measuring, weighing
Polypropylene
Flat
Handle
FDA
82 mm