missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ Polypropylene Test Tube Rack
Brand: VITLAB™ 3191940
This item is not returnable.
View return policy
Specifications
Test Tube Rack | |
White | |
Polypropylene | |
5 Pack | |
75 mm |
5 x 11 | |
Tubes (16 mm) | |
55 | |
5 Pack | |
16 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction