Learn More
Masterflex™ Polypropylene Union Compression Connector Compression Fittings
Compression fittings are ideal when working with any rigid tubing, including tubing made from fluoropolymers, other rigid plastics, and metals.
Brand: Masterflex™ 06381-40
This item is not returnable.
View return policy
Description
- With nut that compresses rigid tubing against fitting body for high-pressure applications
- Withstands higher pressures than barbed fittings
- Forms a secure seal - no leaks, flow restrictions or costly downtime
- Sterilization through ethylene oxide (ETO) only
Specifications
Compression Fitting | |
0.5 in. tubing O.D. | |
10 Pack | |
Ethylene Oxide | |
220 psi |
Polypropylene | |
0°C | |
3/8 in. orifice; 1-13/32 x 15/16 in. | |
Straight | |
100°C |