Learn More
Clark Scientific™ Polystyrene Anti-static Spill Trays
Product Code. 12958837
Click to view available options
Length (Metric):
475 mm
700 mm
Packaging Quantity:
100/Pk.
25/Pk.
Unit Size:
Pack of 100
Pack of 25
Product Code. 12958837
Brand: Clark Scientific™ AT2PK100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Made from anti-static, high impact polystyrene, these spill trays are ideal in most research, scientific, laboratory and other environments.
  • Made from anti-static, high impact polystyrene
  • Sufficient depth to safely contain even the largest liquid spills
  • Wipe clean for repeated use
Specifications
Material Polystyrene
Type Spill Tray
Packaging Quantity 100/Pk.
Length (Metric) 700 mm
Height (Metric) 450 mm