Learn More
Cole-Parmer™ Polystyrene Lazy-L-Spreader
Shop All Cole Parmer Products
Click to view available options
Quantity:
500 Pk.
Unit Size:
Pack of 500
Specifications
Specifications
| Disposable | Single-use |
| Quantity | 500 Pk. |
| Material | Polystyrene |
| Product Type | Spreader |
| Sterility | Sterile |
| For Use With (Application) | Spreaders eliminate flaming for safe, convenient sample plating |