Learn More
Cole-Parmer™ Polystyrene Trans-Boxes container
Shop All Cole Parmer Products
Click to view available options
Height (Metric):
3.5 cm
Unit Size:
Pack of 24
Specifications
Specifications
| Closure Type | Snap Shut Cover |
| Material | Polystyrene |
| For Use With (Application) | Material Handling |
| Product Type | Storage Container |
| Quantity | 24 Pack |
| Length (Metric) | 21 cm |
| Length (English) | 8.25 in. |
| Width (Metric) | 11.4 cm |
| Width (English) | 4.5 in. |
| Height (Metric) | 3.5 cm |
| Show More |