Learn More
Honeywell™ POLYTRIL™ Mix Gloves
Click to view available options
Size:
6
7
8
9
Unit Size:
1 pair
Description
- Black polyamide/cotton knitted glove
- Rough black nitrile coating on palm and fingers
- The knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
- The glove offers maximum comfort and added sturdiness
- The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
- The ventilated back and PU coating optimise the glove's breathability
- The knitted and elasticated wrist gives the glove good support
- The cotton polyamide thread offers excellent comfort and provides the glove with effective properties against cuts
- The nitrile coating at the palm and fingers provides outstanding abrasion resistance and impermeability to oils
- Its black color makes it more soilproof, thus ensuring a longer life
Specifications
Specifications
| Certifications/Compliance | CE CATEGORY : II Intermediary risks In accordance with standards: EN 420-03 Basic standards EN 388-16 Mechanical hazards |
| Description | 10 gauge |
| Coating Material | Polyamide |
| Color | Black |
| Size | 6 |
| Length (Metric) | 25.5 cm |