missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POMK/SGK196 Antibody (OTI2B10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45805
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
POMK/SGK196 Monoclonal specifically detects POMK/SGK196 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| POMK/SGK196 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry 1:150 | |
| FLJ23356, MGC126597, protein kinase-like protein SgK196, Sugen kinase 196 | |
| Mouse | |
| 39.9 kDa | |
| 0.1 mL | |
| Protein Kinase | |
| 84197 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2B10 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| POMK | |
| Human recombinant protein fragment corresponding to amino acids 141-350 of human SGK196 (NP_115613) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction