Learn More
Applied Biosystems™ POP-7™ Polymer for 3730/3730xl DNA Analyzers
Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 x 28 mL
5 x 28 mL
10 x 28 mL
30 x 28 mL
Unit Size:
28mL
Each
Pack of 5
Description
Includes
Each bottle contains enough polymer for 960 samples
- A separation matrix for the 3730 and 3730xl DNA Analyzers
- Customized for DNA sequencing and fragment analysis.
- Pre-formulated to save time and reproducibility by ensuring consistent formulation
- Designed to allow capillaries to be used multiple times for sequencing and fragment analysis applications
- Not suitable for use with ABI PRISM™, 310 and 3100 series genetic analyzers as it causes syringe failure and premature array degradation
- Sufficient for 8,000 to 12,000 samples
Use With
- 3730/3730xl Genetic Analyzer Capillary Array, 50 cm
- 3730 Running Buffer, 10X
- BigDye™ Terminator v1.1 Cycle Sequencing Kit
- BigDye™ Terminator v3.1 Cycle Sequencing Kit
- Hi-Di™ Formamide
Specifications
Specifications
| Accuracy | 98.5% up to 900 bases (for 50 cm capillary), 98.5% up to 700 bases (for 36 cm capillary) |
| Content And Storage | One bottle. Store at 2°C to 8°C. |
| For Use With (Equipment) | 3730xl DNA Analyzer, 3730 DNA Analyzer |
| For Use With (Application) | Standard Sequencing, Fragment Analysis (Denaturing) |
| On Instrument Life Usage | Stability on instrument 7 days, up to 12,000 samples/28 mL |
| Product Line | POP-7 |
| Product Type | Polymer |
| Quantity | 5 x 28 mL |
| Test Time | 1 hr for 36 cm capillary (std. seq.), 2 hrs for 50 cm capillary (long read seq.) |