Learn More
Haldenwanger™ Porcelain Pestle
Click to view available options
Diameter (Metric) Outer:
60mm
180mm
Unit Size:
Each
Description
- Available in a variety of sizes
- Grinding surfaces are unglazed
- Complies with DIN VDE 0335, type C 100
Specifications
Specifications
| Diameter (Metric) Outer | 60mm |
| Product Type | Pestle |
| Material | Porcelain |
| Type | Pestle |