missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ Portable pH/ORP Meters
Brand: Hanna Instruments™ HI9125
Additional Details : Weight : 0.50000kg
Specifications
±0.01 | |
±0.1°C | |
-2.00 to 16.00 | |
±699.9 to ±1999 | |
0.1mV | |
-20.0°C to +120.0°C | |
300 g | |
Stainless Steel temperature probe, BNC connector | |
-20.0°C to +120.0°C | |
Portable pH/ORP Meter |
±0.2 to ±1 | |
Automatic/Manual | |
0.01 | |
±699.9/±1999mV | |
1 or 2 with 5 standard buffer values (pH 4.01, 6.86, 7.01, 9.18, 10.01) | |
0.1°C | |
Stainless Steel Temperature Probe | |
-2.00 to 16.00 pH | |
1.5V | |
200 hr. |