Learn More
Brady™ Post and Cement Bases
Click to view available options
Color:
Black and Yellow Striped
Red
Red on White
White
Unit Size:
1 pack
Description
- Ideal for controlling and directing foot traffic.
- Suitable for warehouses, factories, offices etc.
- Chains and ribbons can be attached to the sturdy 900 mm posts.
- Includes Post and Cement Base.
Specifications
Specifications
| Alternate Part Number | BRADYPOST & CEMENT BASE YEL/BLK PK4 |
| Diameter (Metric) Outer | 40 mm |
| Height (English) | 35.43 in. |
| Height (Metric) | 900 mm |
| Packaging Quantity | 4 Pk. |
| Color | Black and Yellow Striped |
| Type | Post with Cement Base |