Learn More
Brady™ Post Top
Click to view available options
Color:
Black
Red
White
Unit Size:
Each
Description
- Ideal for controlling and directing foot traffic.
- Suitable for warehouses, factories, offices etc.
- Chains and ribbons can be attached to the sturdy 900 mm posts.
- Top only.
Specifications
Specifications
| Diameter (Metric) Outer | 60 mm |
| Packaging Quantity | 1 Ea. |
| Color | White |
| Type | Post Top |