missing translation for 'onlineSavingsMsg'
Learn More
Potassium 1-Boc-indole-2-trifluoroborate, 95%, Thermo Scientific™ Product Code.: 15485187

Potassium 1-Boc-indole-2-trifluoroborate, 95%, Thermo Scientific™

Product Code. 15485187
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15485187

Brand: Thermo Scientific Alfa Aesar H32467.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 945493-51-6
Molecular Formula C13H14BF3KNO2
Molecular Weight (g/mol) 323.164
MDL Number MFCD12912217
InChI Key IIPRPASPOXMUTF-UHFFFAOYSA-N
Synonym potassium 1-tert-butoxycarbonly-1h-indole-2-yl trifluoroborate, potassium 1-boc-1h-indole-2-trifluoroborate, potassium 1-boc-1h-indol-2-yl trifluoroborate, potassium 1-tert-butoxycarbonyl indol-2-yl trifluoroboranuide, potassium 1-tert-butoxycarbonyl-1h-indole-2-yl trifluoroborate, potassium 1-tert-butoxycarbonyl-1h-indole-2-trifluoroborate, potassium 1-tert-butoxycarbonyl-1h-indol-2-yl trifluoroborate, potassium 1-tert-butoxycarbonyl-1h-indol-2-yl trifluoro borate 1-
PubChem CID 23664789
IUPAC Name potassium;trifluoro-[1-[(2-methylpropan-2-yl)oxycarbonyl]indol-2-yl]boranuide
SMILES [B-](C1=CC2=CC=CC=C2N1C(=O)OC(C)(C)C)(F)(F)F.[K+]

Specifications

Quantity 250 mg
Formula Weight 323.17
Percent Purity 95%
Chemical Name or Material Potassium 1-Boc-indole-2-trifluoroborate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberCB388-1 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium 1-Boc-indole-2-trifluoroborate, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.