missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride, puriss. p.a., Honeywell Fluka™

Puriss. p.a., Reag. ISO, Reag. Ph. Eur., 99.5-100.5%

33.70 EUR - 992.87 EUR

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]
View More Specs

Products 7
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15633320
View Documents
Honeywell-Fluka
31248-100G
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15673320
View Documents
Honeywell-Fluka
31248-500G
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15643320
View Documents
Honeywell-Fluka
31248-1KG
1 kg
73.49 EUR
1kg
Please to purchase this item. Need a web account? Register with us today!
15683320
View Documents
Honeywell-Fluka
31248-5KG
5 kg
308.81 EUR
5kg
Please to purchase this item. Need a web account? Register with us today!
15693320
View Documents
Honeywell-Fluka
31248-6X1KG
6 x 1 kg
333.83 EUR
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15663320
View Documents
Honeywell-Fluka
31248-4X5KG
4 x 5 kg
992.87 EUR
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
15653320
View Documents
Honeywell Fluka
31248-25KG
25 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7447-40-7
74.55
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
4873
[Cl-].[K+]
ClK
MFCD00011360
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
CHEBI:32588

Specifications

7447-40-7
KCl
NONH for all modes of transport
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride
4873
74.55g/mol
puriss. p.a.
Potassium Chloride, Reag. Ph. Eur. reag. ISO
ClK
MFCD00011360
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
[Cl-].[K+]
74.55
CHEBI:32588
99.5 to 100.5%
1.98g/mL at 25°C
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-211-8

missing translation for 'rtecsNumber' : TS8050000

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.