missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 84μS, Chem-Lab

Product Code. 11910339
Click to view available options
Quantity:
500 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11910339

Brand: Chem-Lab CL03.1150.0500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Calibrates conductivity measurement equipment. Potassium Chloride Conductivity Standard, 84μS, Chem-Lab is precisely concentrated to a specific LF electrolyte conductivity level in order to calibrate and verify the performance of conductivity meters and probes.

Use of conductivity standard ensures that instruments are properly calibrated for accurate readings. By placing probes into a solution with an identified transfer ability, the equipment settings can be adjusted prior to testing. KCl solutions are used to calibrate conductivity instruments that read directly in microsiemens or millisiemens. Potassium chloride is a very stable salt and is an international calibration standard for conductivity measurement.

Specifications

CAS 7447-40-7
Odor Odorless
Density 1.00g/mL
Conductivity 84.0μS at 25°C,
75.8μS at 20°C (±1%)
Physical Form Liquid
Molecular Formula ClK
MDL Number MFCD00011360
Color Colorless
pH ±5
For Use With (Equipment) Conductivity Meter
Solubility Soluble
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES [Cl-].[K+]
IUPAC Name potassium;chloride
Molecular Weight (g/mol) 74.55
PubChem CID 4873
Quantity 500 mL
ChEBI CHEBI:32588
Formula Weight 74.56g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.