Learn More
Potassium hydrogen iodate, primary standard, ACS, 99.95-100.05%, Thermo Scientific Chemicals
Maximum level of impurities: Insoluble matter 0.01%, SO4 0.005%, Fe 0.001%, Cl 0.02%, Heavy Metals (as Pb) 0.001%
81.51 EUR - 264.32 EUR
Chemical Identifiers
CAS | 13455-24-8 |
---|---|
Molecular Formula | HI2KO6 |
Molecular Weight (g/mol) | 389.909 |
MDL Number | MFCD00011400 |
InChI Key | ACAYDTMSDROWHW-UHFFFAOYSA-M |
PubChem CID | 23700942 |
IUPAC Name | potassium;iodic acid;iodate |
SMILES | OI(=O)=O.[O-]I(=O)=O.[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11344987
|
Thermo Scientific Alfa Aesar
041986.14 |
25 g |
81.51 EUR
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11334987
|
Thermo Scientific Alfa Aesar
041986.22 |
100 g |
264.32 EUR
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium hydrogen iodate acts as an oxidizing agent. Further, it serves as an intermediate for the synthesis of potassium iodate and also used as an analysis reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13455-24-8 | |
389.909 | |
ACAYDTMSDROWHW-UHFFFAOYSA-M | |
potassium;iodic acid;iodate |
HI2KO6 | |
MFCD00011400 | |
23700942 | |
OI(=O)=O.[O-]I(=O)=O.[K+] |
Specifications
13455-24-8 | |
99.95-100.05% | |
MFCD00011400 | |
OI(=O)=O.[O-]I(=O)=O.[K+] | |
389.909 | |
389.91 | |
Soluble in water | |
Potassium hydrogen iodate, Primary Standard |
White | |
HI2KO6 | |
ACAYDTMSDROWHW-UHFFFAOYSA-M | |
potassium;iodic acid;iodate | |
23700942 | |
ACS | |
Powder |
Safety and Handling
Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H272-H314-H335
missing translation for 'dotInformation' : DOT Class: 5.1, Packaging Group: II
missing translation for 'einecsNumber' : 236-650-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only