missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 0.1M, Reagecon™
BIOPHARMA

Product Code. 11556833
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Used to determine KOH content and in acid determination in wine. Potassium Hydroxide, 0.1M, Reagecon™ is a ready-to-use analytical volumetric solution with 5.61g/L HCl in water for use as a laboratory reagent. It is also suitable for use in the wine industry.

Potassium Hydroxide is a prototypical strong base that is corrosive and reactive toward acids. It is soluble in water, forming an electrolyte solution. This is an analytical volumetric solution used in manual and automated titrations. This solution meets all test methods included in the Compendium of International Methods of Wine and Must Analysis, and can be used in determination of tartaric acid in wine.

Specifications

pH 13
Color Colorless
Boiling Point 100°C
CAS 1310-58-3, 7732-18-5
Molecular Formula HKO
Concentration 0.1M
MDL Number MFCD00003553
Density 1.00g/cm3
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Quantity 1 L
SMILES [OH-].[K+]
IUPAC Name potassium;hydroxide
Molecular Weight (g/mol) 56.11
PubChem CID 14797
ChEBI CHEBI:32035
Formula Weight 56.106g/mol
Concentration or Composition (by Analyte or Components) 5.61g/L KOH in water
Normality 0.1N
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • POTASSIUM HYDROXIDE 0.1N (0.1M)
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Potassium Hydroxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'