missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Nitrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
5 kg
Description
KNO3, CAS Number-7757-79-1, nitre, saltpeter, nitric acid potassium salt, kaliumnitrat, nitrate of potash, vicknite, nitric acid, potassium salt, kalii nitras, potassium nitrate, saltpetre, 500g, 400 deg.C, CHEBI:63043, White, 101.1g/mol, FGIUAXJPYTZDNR-UHFFFAOYSA-N, potassium;nitrate, 334 deg.C
Specifications
Specifications
Boiling Point | 400°C |
Melting Point | 334°C |
Color | White |
pH | 6 to 8 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 101.1g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Potassium nitrate
- Warning
- Oxidising solid Category 3
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction