missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Potassium permanganate, 99+%, for analysis

Product Code. 10219843
Click to view available options
Quantity:
25 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-64-7
Molecular Formula KMnO4
MDL Number MFCD00011364
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
Synonym argucide, cairox, solo san soo, insta-perm, condy's crystals, walko tablets, potassium permanganate, chameleon mineral, algae-k, permanganate of potash
PubChem CID 516875

Specifications

Color Green to Black
Physical Form Crystalline Powder and/or Crystals
Quantity 5 kg
Assay Percent Range 99+%
Linear Formula KMnO4
Fieser 01,942; 02,348; 04,412; 05,562; 08,416; 09,388; 10,330; 11,440; 12,413; 13,258; 14,267; 15,273; 16,283
Merck Index 15, 7776
SMILES [K+].[O-][Mn](=O)(=O)=O
IUPAC Name potassium manganesoylolate
Molecular Weight (g/mol) 158.03
Formula Weight 158.04
Percent Purity ≥99.0%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Potassium permanganate, For analysis
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'