missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium permanganate, Puriss., meets analytical spec. of BP, Ph. Eur., USP, 99 to 100.5%, Honeywell Fluka™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
222.17 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
25 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 2.5 kg |
UN Number | 1490 |
Formula Weight | 158.03g/mol |
Percent Purity | 99 to 100.5% |
Grade | puriss. |
Chemical Name or Material | Potassium Permanganate, Meets analytical specification of Ph. Eur.BP, USP |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'