missing translation for 'onlineSavingsMsg'
Learn More

Potassium Standard for ICP, ICP-MS 10,000 μg/mL (10,000 ppm) in 2-5% Nitric Acid (HNO3), Reagecon
BIOPHARMA

Manufactured from ultra-pure metal (99.999%) tested by an ISO17025 Accredited Test Method (INAB Ref: 264T) and raw materials assayed by titration and ICP-MS prior to manufacture.

Brand:  Reagecon PK4B2

Product Code. 11418999

  • 272.00 EUR / 250mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • ICP, ICP-MS Reference Standard, ICP, ICP-MS Reference Materia
  • ISO/IEC 17025 for the testing of ICP/ICP-MS Standards using ICP-MS (INAB Ref: 264T)
  • ISO/IEC 17025 for calibration of laboratory balances (INAB Ref: 265C)
  • ISO/IEC 17025 for temperature calibration in the range of -196 - +1200°C (INAB Ref: 265C)
  • ISO/IEC 17025 for density measurement (INAB Ref: 264T) using Oscillating U-Tube and Bingham Pycnometry
  • ISO/IEC 17025 for certain complexometric titrations (INAB Ref: 264T)
  • GHS/OSHA compliant, REACH etc.
  • This product is produced in a highly controlled cleanroom (ISO 7) environment, using ultra-pure water (specially treated for the production of mass spectroscopy standards), triple distilled acids and triple leached pre-cleaned bottles.
  • The titrimetry and spectroscopy are controlled and calibrated using separate Certified Reference Materials (CRM’s), so the product comes with two layers of traceability including NIST Standards where available.
  • All of the uncertainties of measurement for this product are calculated according to Eurachem/CITAC guidelines and reported as expanded uncertainties at the 95% confidence level.
Specifications

Specifications

Matrix: 2-5% HNO3
HDPE Triple-leached
Potassium
Liquid
10,000 µg/mL (10,000 ppm)
250 mL
ICP, ICP-MS, AA, IC, XRF and other cation measurement techniques
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.