Learn More
WTW™ Potassium Test Kits
Product Code. 11791113
Click to view available options
No. of Tests:
25
Unit Size:
Pack of 25
Product Code. 11791113
Brand: WTW™ 250407
Specifications
Analytes Chromosulfuric Acid oxidation
For Use With (Application) For drinking water, ground water, surface water, well water, curative water, soils
For Use With (Equipment) PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, spectroFlex, Photoflex photometers
Includes Certificates
No. of Tests 25
Test Method Photometric
compliance-icons
Product Identifier
  • potassium cell test method
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P313-Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : ethylenedinitrilotetraacetic acid, tetrasodium salt