missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Potassium thiocyanate, 99%, for biochemistry

Product Code. 12615137
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
Molecular Weight (g/mol) 97.18
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Melting Point 170.0°C to 179.0°C
Quantity 1 kg
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula KSCN
Merck Index 15,7807
Solubility Information Solubility in water: 2170g/L (20°C). Other solubilities: soluble in alcohol and acetone
Formula Weight 97.18
Percent Purity 99%
Grade Biochemical
Packaging Plastic bottle
Specific Gravity 1.886
Density 1.8860g/mL
Chemical Name or Material Potassium thiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'