Learn More
Memmert™ Potential-Free Contact
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP1400eco
HPP2200eco
HPP260eco
HPP410eco
HPP750eco
ICO105
ICO150
ICO240
ICO50
IPP1060eco
IPP110eco
IPP1400eco
IPP2200eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Description
- Accessory
- For combination error message (e.g. supply failure,sensor fault, fuse)
Specifications
Specifications
| Description | Combination error message |
| For Use With (Equipment) | IPP110eco |