Learn More
Invitrogen™ Power Blotter Pre-cut Membranes and Filters, PVDF mini
Precut, preassembled PVDF membrane and filter paper sandwiches for midi sized gels
281.00 EUR
Specifications
Material PVDF
Pore Size 0.45 μm
Shipping Condition Room Temperature
View More Specs
Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15868502
View Documents Promotion Details
Invitrogen™
PB9220
281.00 EUR
Pack of 20
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
Description
Description
The Invitrogen Power Blotter Pre-cut Membranes and Filters are designed for transferring proteins from gels using the Invitrogen Power Blotter System.
Features:
  • Membranes and filters allow for high efficiency semi-dry transfer of proteins in less than 10 minutes when combined with the Power Blotter 1-Step Transfer Buffer
  • PVDF Membranes (0.45μM pore size, low fluorescence) have a higher binding capacity than nitrocellulose and are manufactured especially for protein transfer and western blot applications
    • Compatible with commonly used detection methods such as staining, immunodetection, or fluorescence.
    • 240 μg/cm2 protein binding affinity blocks easily and is compatible with most organic solvents, acids and mild bases.  Proteins bind to the membrane as hydrophobic interactions
    • Membrane has lower background levels and increased sensitivity for flourescent probing than regular PVDF or nitrocellulose membranes
  • Convenient— Pre-cut sheets are ready to use, meaning no need to cut your own membranes, minimizing damage and contamination
  • Validated – When combined, membrane and filter provide the thickness and buffer capacity needed for optimal performance
Specifications
Specifications
PVDF
0.45 μm
Room Temperature
Videos
SDS
Documents
Documents
Product Certifications
For Research Use Only. Not for use in diagnostic procedures.