Learn More
Invitrogen™ Power Blotter Pre-cut Membranes and Filters, PVDF mini
Precut, preassembled PVDF membrane and filter paper sandwiches for midi sized gels
281.00 EUR
Description
The Invitrogen Power Blotter Pre-cut Membranes and Filters are designed for transferring proteins from gels using the Invitrogen Power Blotter System.
Features:
- Membranes and filters allow for high efficiency semi-dry transfer of proteins in less than 10 minutes when combined with the Power Blotter 1-Step Transfer Buffer
- PVDF Membranes (0.45μM pore size, low fluorescence) have a higher binding capacity than nitrocellulose and are manufactured especially for protein transfer and western blot applications
- Compatible with commonly used detection methods such as staining, immunodetection, or fluorescence.
- 240 μg/cm2 protein binding affinity blocks easily and is compatible with most organic solvents, acids and mild bases. Proteins bind to the membrane as hydrophobic interactions
- Membrane has lower background levels and increased sensitivity for flourescent probing than regular PVDF or nitrocellulose membranes
- Convenient— Pre-cut sheets are ready to use, meaning no need to cut your own membranes, minimizing damage and contamination
- Validated – When combined, membrane and filter provide the thickness and buffer capacity needed for optimal performance
Specifications
PVDF | |
0.45 μm | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.